missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Alfa Aesar™ N-Boc-1-formyl-L-tryptophan, 98%

Manufacturer:  Alfa Aesar™ H6349906

 View more versions of this product

Catalog No. AAH6349906



Specifications

Specifications

N-Boc-1-formyl-L-tryptophan
C17H20N2O5
4299614
IHXHBYFWSOYYTR-ZDUSSCGKSA-N
(2S)-3-(1-formylindol-3-yl)-2-[(2-methylpropan-2-yl)oxycarbonylamino]propanoic acid
7017963
Solid
100°C (decomposition)
Light Sensitive
47355-10-2
MFCD00065992
boc-trp for-oh, n-tert-butoxycarbonyl-n'-formyl-l-tryptophan, chembl63415, nalpha-tert-butoxycarbonyl-n1-formyl-l-tryptophan, s-2-tert-butoxycarbonyl amino-3-1-formyl-1h-indol-3-yl propanoic acid, 2s-2-tert-butoxycarbonyl amino-3-1-formylindol-3-yl propanoic acid, boc-trp cho-oh, pubchem12191, t-boc-trp cho-oh, nalpha-boc-n1-formyl-l-tryptophan
CC(C)(C)OC(=O)NC(CC1=CN(C2=CC=CC=C21)C=O)C(=O)O
332.356
332.35
98%
5g

Chemical Identifiers

47355-10-2
332.356
IHXHBYFWSOYYTR-ZDUSSCGKSA-N
7017963
CC(C)(C)OC(=O)NC(CC1=CN(C2=CC=CC=C21)C=O)C(=O)O
C17H20N2O5
MFCD00065992
boc-trp for-oh, n-tert-butoxycarbonyl-n'-formyl-l-tryptophan, chembl63415, nalpha-tert-butoxycarbonyl-n1-formyl-l-tryptophan, s-2-tert-butoxycarbonyl amino-3-1-formyl-1h-indol-3-yl propanoic acid, 2s-2-tert-butoxycarbonyl amino-3-1-formylindol-3-yl propanoic acid, boc-trp cho-oh, pubchem12191, t-boc-trp cho-oh, nalpha-boc-n1-formyl-l-tryptophan
(2S)-3-(1-formylindol-3-yl)-2-[(2-methylpropan-2-yl)oxycarbonylamino]propanoic acid
Safety and Handling

Safety and Handling

storageNote1 : -30° to -10°C

Documents