Please login to your online account to display your discounted pricing

Alfa Aesar™ N-Boc-1-formyl-L-tryptophan, 98%

Chemical Identifiers

Chemical Name or Material N-Boc-1-formyl-L-tryptophan
CAS 47355-10-2
Formula Weight 332.35
MDL Number MFCD00065992
Beilstein 4299614
Synonym Boc-Trp(For)-OH; (S)-2-(Boc-amino)-3-(1-formyl-3-indolyl)propionic acid
View More Specs
Products
Catalog Number Mfr. No. Quantity Price Quantity    

AAH6349906

 
alfa aesar™
H6349906
5g CALL N/A N/A
 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

AAH6349914

 
alfa aesar™
H6349914
25g CALL N/A N/A
 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

AAH6349922

 
alfa aesar™
H6349922
100g CALL N/A N/A
 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
Description & Specifications

Specifications

Formula Weight 332.35
Melting Point 100°C (dec.)
Chemical Name or Material N-Boc-1-formyl-L-tryptophan
Physical Form Solid
Assay 98%
MDL Number MFCD00065992
CAS 47355-10-2
Beilstein 4299614
Synonym Boc-Trp(For)-OH; (S)-2-(Boc-amino)-3-(1-formyl-3-indolyl)propionic acid
Quantity 5g
Sensitivity Light Sensitive
Storage Note 1 -30° to -10°C
Safety and Handling

-30° to -10°C