Please login to your online account to display your discounted pricing

N-boc-1,6-hexanediamine Hydrochloride, ACROS Organics™


Quantity 1g
Packaging Glass bottle

Manufacturer: thermo fisher scientific  AC381490010

 View more versions of this product

Catalog No. AC381490010

 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.


Description & Specifications

Chemical Identifiers

Assay ≥94%
Formula Weight 252.79
Physical Form Crystalline Powder
Color White to cream
Quantity 1g
Melting Point 155° to 165°C
Linear Formula NH2(CH2)6NHCOOC(CH3)3·HCl
Molecular Formula C11H24N2O2·HCl
CAS 65915-94-8
Health Hazard 3 GHS P Statement:
Avoid breathing dust/fume/gas/mist/vapors/spray.
IF ON SKIN: Wash with plenty of soap and water.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.
MDL Number MFCD00039072
Infrared Spectrum Authentic
Synonym N-tert-Butoxycarbonyl-1, 6-hexanediamine hydrochloride
Packaging Glass bottle
Chemical Name or Material N-BOC-1, 6-hexanediamine hydrochloride
Health Hazard 2 GHS H Statement:
May cause respiratory irritation.
Causes serious eye irritation.
Causes skin irritation.

Specifications

Chemical Name or Material N-BOC-1, 6-hexanediamine hydrochloride
Assay ≥94%
Molecular Formula C11H24N2O2·HCl
Linear Formula NH2(CH2)6NHCOOC(CH3)3·HCl
Formula Weight 252.79
MDL Number MFCD00039072
Synonym N-tert-Butoxycarbonyl-1, 6-hexanediamine hydrochloride
Accessories

GHS P Statement:
Avoid breathing dust/fume/gas/mist/vapors/spray.
IF ON SKIN: Wash with plenty of soap and water.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.

GHS H Statement:
May cause respiratory irritation.
Causes serious eye irritation.
Causes skin irritation.

SDS & Safety Documents