missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Please login to your online account to display your discounted pricing
N-Boc-1,6-diaminohexane hydrochloride, 98%, Thermo Scientific Chemicals
Supplier: Thermo Scientific Chemicals L1948706
Description
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Specifications
∽164°C (decomposition) | |
65915-94-8 | |
MFCD00039072 | |
n-boc-1,6-hexanediamine hydrochloride, tert-butyl 6-aminohexyl carbamate hydrochloride, n-boc-1,6-diamino-hexane hydrochloride, tert-butyl n-6-aminohexyl carbamate hydrochloride, n-6-aminohexyl tert-butoxy carboxamide, chloride, tert-butyl 6-aminohexyl carbamate monohydrochloride, tert-butyl 6-aminohexyl carbamate hcl, acmc-1b5cm, n-1-boc-1,6-diaminohexane . hcl, n-boc-1,6-hexanediamine hcl | |
[H+].[Cl-].CC(C)(C)OC(=O)NCCCCCCN | |
252.78 | |
252.79 | |
5 g |
N-Boc-1,6-diaminohexane hydrochloride | |
C11H25ClN2O2 | |
3631740 | |
JSBWQIZQJOQPFN-UHFFFAOYSA-N | |
tert-butyl N-(6-aminohexyl)carbamate;hydrochloride | |
3017638 | |
98% | |
Hygroscopic |
Chemical Identifiers
65915-94-8 | |
252.78 | |
JSBWQIZQJOQPFN-UHFFFAOYSA-N | |
3017638 | |
[H+].[Cl-].CC(C)(C)OC(=O)NCCCCCCN |
C11H25ClN2O2 | |
MFCD00039072 | |
n-boc-1,6-hexanediamine hydrochloride, tert-butyl 6-aminohexyl carbamate hydrochloride, n-boc-1,6-diamino-hexane hydrochloride, tert-butyl n-6-aminohexyl carbamate hydrochloride, n-6-aminohexyl tert-butoxy carboxamide, chloride, tert-butyl 6-aminohexyl carbamate monohydrochloride, tert-butyl 6-aminohexyl carbamate hcl, acmc-1b5cm, n-1-boc-1,6-diaminohexane . hcl, n-boc-1,6-hexanediamine hcl | |
tert-butyl N-(6-aminohexyl)carbamate;hydrochloride |
Safety and Handling
P261-P264b-P271-P280-P302+P352-P304+P340-P305+P351+P338-P312-P332+P313-P362-P501c
H315-H319-H335
EINECSNumber : 265-976-4
TSCA : No
Recommended Storage : Ambient temperatures
RUO – Research Use Only