Please login to your online account to display your discounted pricing

N-boc-1,3-Propanediamine Hydrochloride 97%, ACROS Organics™


Quantity 5g
Packaging Glass bottle

Manufacturer: thermo fisher scientific  AC380580050

 View more versions of this product

Catalog No. AC380580050

 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.


Description & Specifications

Chemical Identifiers

Assay ≥96.0%
Formula Weight 210.71
Physical Form Crystalline Powder
Quantity 5g
Color White
CAS Max % 100.0
CAS Min % 96.0
Synonym N-tert-Butoxycarbonyl-1, 3-propanediamine hydrochloride
Assay Percent Range 97%
Health Hazard 1 Warning
Molecular Formula C8H18N2O2·HCl
Infrared Spectrum Authentic
Packaging Glass bottle
Linear Formula (CH3)3CHOC(O)NH(CH2)3NH2·HCl
Chemical Name or Material N-BOC-1, 3-propanediamine hydrochloride
CAS 127346-48-9
Health Hazard 3 GHS P Statement:
Avoid breathing dust/fume/gas/mist/vapors/spray.
IF ON SKIN: Wash with plenty of soap and water.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.
MDL Number MFCD01076598
Health Hazard 2 GHS H Statement:
Causes serious eye irritation.
Causes skin irritation.
May cause respiratory irritation.

Specifications

Chemical Name or Material N-BOC-1, 3-propanediamine hydrochloride
CAS 127346-48-9
CAS Min % 96.0
CAS Max % 100.0
Assay ≥96.0%
Assay Percent Range 97%
Molecular Formula C8H18N2O2·HCl
Linear Formula (CH3)3CHOC(O)NH(CH2)3NH2·HCl
Formula Weight 210.71
MDL Number MFCD01076598
Synonym N-tert-Butoxycarbonyl-1, 3-propanediamine hydrochloride
Accessories

Warning

GHS P Statement:
Avoid breathing dust/fume/gas/mist/vapors/spray.
IF ON SKIN: Wash with plenty of soap and water.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.

GHS H Statement:
Causes serious eye irritation.
Causes skin irritation.
May cause respiratory irritation.

SDS & Safety Documents