missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

N-boc-1,3-Propanediamine Hydrochloride 97%, ACROS Organics™

Chemical Identifiers

CAS 127346-48-9
CAS Min % 96.0
CAS Max % 100.0
Assay Percent Range 97%
Molecular Formula C8H18N2O2·HCl
Linear Formula (CH3)3CHOC(O)NH(CH2)3NH2·HCl
Formula Weight 210.71
MDL Number MFCD01076598
Synonym N-tert-Butoxycarbonyl-1, 3-propanediamine hydrochloride
Chemical Name or Material N-BOC-1, 3-propanediamine hydrochloride
View More Specs


Products ${productFamilyLength}
Catalog Number Mfr. No. Packaging Quantity Price Quantity  
Catalog Number Mfr. No. Packaging Quantity Price Quantity  
AC380580010 SDS Fisher Scientific Acros
AC380580010
Glass bottle 1g N/A N/A  
Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
AC380580050 SDS Thermo Fisher Scientific
AC380580050
Glass bottle 5g N/A N/A  
Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
Specifications Description & Specifications

Chemical Identifiers

127346-48-9
100.0
C8H18N2O2·HCl
210.71
N-tert-Butoxycarbonyl-1, 3-propanediamine hydrochloride
≥96.0%
96.0
97%
(CH3)3CHOC(O)NH(CH2)3NH2·HCl
MFCD01076598
N-BOC-1, 3-propanediamine hydrochloride

Specifications

127346-48-9
100.0
Glass bottle
97%
(CH3)3CHOC(O)NH(CH2)3NH2·HCl
MFCD01076598
N-tert-Butoxycarbonyl-1, 3-propanediamine hydrochloride
N-BOC-1, 3-propanediamine hydrochloride
96.0
Authentic
Crystalline Powder
C8H18N2O2·HCl
210.71
White
1g
≥96.0%

Safety and Handling

GHS H Statement:
Causes serious eye irritation.
Causes skin irritation.
May cause respiratory irritation.

GHS P Statement:
Avoid breathing dust/fume/gas/mist/vapors/spray.
IF ON SKIN: Wash with plenty of soap and water.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.

Warning