Please login to your online account to display your discounted pricing

Alfa Aesar™ N-Boc-1,3-diaminopropane hydrochloride, 98%

$57.86 - $146.16

Chemical Identifiers

Chemical Name or Material N-Boc-1,3-diaminopropane hydrochloride
CAS 127346-48-9
Molecular Formula (CH3)3COCONHCH2CH2CH2NH2·HCl
Formula Weight 210.71
MDL Number MFCD01076598
Beilstein 3627894
Synonym N-Boc-1,3-propanediamine hydrochloride; N-tert-Butoxycarbonyl-1,3-diaminopropane hydrochloride
View More Specs
Products
Catalog Number Mfr. No. Quantity Price Quantity    

AAL1948603

  SDS
alfa aesar™
L1948603
1g Each for $146.16

AAL19486MD

  SDS
alfa aesar™
L19486MD
250mg Each for $57.86
Description & Specifications

Specifications

Formula Weight 210.71
Synonym N-Boc-1,3-propanediamine hydrochloride; N-tert-Butoxycarbonyl-1,3-diaminopropane hydrochloride
Chemical Name or Material N-Boc-1,3-diaminopropane hydrochloride
CAS 127346-48-9
MDL Number MFCD01076598
Molecular Formula (CH3)3COCONHCH2CH2CH2NH2·HCl
Assay 98%
Beilstein 3627894
Melting Point 159° to 161°C
Quantity 1g