missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing
N-BOC-1,2-diaminoethane, 95%, Maybridge™
Chemical Identifiers
CAS
57260-73-8
Molecular Formula
C7H16N2O2
Molecular Weight (g/mol)
160.217
MDL Number
MFCD00191871
InChI Key
AOCSUUGBCMTKJH-UHFFFAOYSA-N
Synonym
mono-n-t-boc-eda, n-boc-1,2-diaminoethane, n-boc-diaminoethane, n-boc-ethane-1,2-diamine, n-boc-ethylenediamine, n-t-boc-ethylenediamine, n-tert-boc-ethylenediamine, tert-butyl 2-aminoethyl carbamate, tert-butyl 2-aminoethylcarbamate, tert-butyl n-2-aminoethyl carbamate
PubChem CID
187201
IUPAC Name
tert-butyl N-(2-aminoethyl)carbamate
SMILES
CC(C)(C)OC(=O)NCCN
View More Specs
Products
1
Catalog Number
Mfr. No.
Quantity
Packaging
Price
Quantity
AW00372EA
SDS
View Documents
Fisher Scientific Company
AW00372EA
10g
Amber Glass Bottle
N/A
N/A
Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
Description
Specifications
Chemical Identifiers
CAS
57260-73-8
Molecular Weight (g/mol)
160.217
InChI Key
AOCSUUGBCMTKJH-UHFFFAOYSA-N
PubChem CID
187201
SMILES
CC(C)(C)OC(=O)NCCN
Molecular Formula
C7H16N2O2
MDL Number
MFCD00191871
Synonym
mono-n-t-boc-eda, n-boc-1,2-diaminoethane, n-boc-diaminoethane, n-boc-ethane-1,2-diamine, n-boc-ethylenediamine, n-t-boc-ethylenediamine, n-tert-boc-ethylenediamine, tert-butyl 2-aminoethyl carbamate, tert-butyl 2-aminoethylcarbamate, tert-butyl n-2-aminoethyl carbamate
IUPAC Name
tert-butyl N-(2-aminoethyl)carbamate
Specifications
Density
1.016g/mL
Quantity
10g
CAS Min %
94.5
Assay Percent Range
95%
Linear Formula
(CH3 )3 COC(O)NH(CH2 )2 NH2
Synonym
mono-n-t-boc-eda, n-boc-1,2-diaminoethane, n-boc-diaminoethane, n-boc-ethane-1,2-diamine, n-boc-ethylenediamine, n-t-boc-ethylenediamine, n-tert-boc-ethylenediamine, tert-butyl 2-aminoethyl carbamate, tert-butyl 2-aminoethylcarbamate, tert-butyl n-2-aminoethyl carbamate
SMILES
CC(C)(C)OC(=O)NCCN
Molecular Weight (g/mol)
160.217
Formula Weight
160.22
Chemical Name or Material
Tert-butyl N-(2-aminoethyl)carbamate
Packaging
Amber Glass Bottle
CAS
57260-73-8
CAS Max %
100.0
Molecular Formula
C7H16N2O2
MDL Number
MFCD00191871
InChI Key
AOCSUUGBCMTKJH-UHFFFAOYSA-N
IUPAC Name
tert-butyl N-(2-aminoethyl)carbamate
PubChem CID
187201
Percent Purity
95%
Safety and Handling
Safety and Handling
GHS H Statement: Causes severe skin burns and eye damage. Causes serious eye damage.
GHS P Statement: IF SWALLOWED: rinse mouth. Do NOT induce vomiting. Wear eye protection/face protection. IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing. Immediately call a POISON CENTER or doctor/physician.WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.
GHS Signal Word: Danger
SDS