Learn More
N-Boc-1,2,5,6-tetrahydropyridine-4-boronic acid pinacol ester, 95%, Thermo Scientific Chemicals
Supplier: Thermo Scientific Chemicals H6619306
Description
Reagent used for suzuki-Miyaura cross-coupling using palladium phosphine catalyst, palladium-catalyzed ligand-controlled regioselective Suzuki coupling, wrenchnolol derivative optimized for gene activation in cells. Also used in Preparation of several enzymatic inhibitors and receptor ligands, orally active anaplastic lymphoma kinase inhibitors and oxazolecarboxamides as diacylglycerol acyltransferase-1 inhibitors for treatment of obesity and diabetes.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
ApplicationsReagent used for suzuki-Miyaura cross-coupling using palladium phosphine catalyst, palladium-catalyzed ligand-controlled regioselective Suzuki coupling, wrenchnolol derivative optimized for gene activation in cells. Also used in Preparation of several enzymatic inhibitors and receptor ligands, orally active anaplastic lymphoma kinase inhibitors and oxazolecarboxamides as diacylglycerol acyltransferase-1 inhibitors for treatment of obesity and diabetes.
Solubility
Soluble in most organic solvents. Insoluble in water.
Notes
Store in cool, dry place in tightly closed container. With adequate ventilation. Store away from oxidizing agent.
Specifications
N-Boc-1,2,5,6-tetrahydropyridine-4-boronic acid pinacol ester | |
C16H28BNO4 | |
5 g | |
Soluble in most organic solvents. Insoluble in water. | |
CC(C)(C)OC(=O)N1CCC(=CC1)B1OC(C)(C)C(C)(C)O1 | |
309.21 | |
327.23 | |
Solid |
286961-14-6 | |
MFCD03840345 | |
n-boc-1,2,5,6-tetrahydropyridine-4-boronic acid pinacol ester, tert-butyl 4-4,4,5,5-tetramethyl-1,3,2-dioxaborolan-2-yl-5,6-dihydropyridine-1 2h-carboxylate, 1-n-boc-4-4,4,5,5-tetramethyl-1,3,2 dioxaborolan-2-yl-3,6-dihydro-2h-pyridine, n-boc-1,2,3,6-tetrahydropyridine-4-boronic acid pinacol ester, tert-butyl 4-4,4,5,5-tetramethyl-1,3,2-dioxaborolan-2-yl-3,6-dihydro-2h-pyridine-1-carboxylate, tert-butyl 5,6-dihydro-4-4,4,5,5-tetramethyl-1,3,2-dioxaborolan-2-yl pyridine-1 2h-carboxylate, 3,6-dihydro-2h-pyridine-1-n-boc-4-boronic acid pinacol ester, n-tert-butoxycarbonyl-1,2,3,6-tetrahydropyridine-4-boronic acid pinacol ester, 1-boc-4-4,4,5,5-tetramethyl-1,3,2 dioxaborolan-2-yl-3,6-dihydro-2h-pyridine, tert-butyl 4-tetramethyl-1,3,2-dioxaborolan-2-yl-1,2,3,6-tetrahydropyridine-1-carboxylate | |
VVDCRJGWILREQH-UHFFFAOYSA-N | |
tert-butyl 4-(4,4,5,5-tetramethyl-1,3,2-dioxaborolan-2-yl)-3,6-dihydro-2H-pyridine-1-carboxylate | |
4642098 | |
95% |
Chemical Identifiers
286961-14-6 | |
309.21 | |
VVDCRJGWILREQH-UHFFFAOYSA-N | |
4642098 | |
CC(C)(C)OC(=O)N1CCC(=CC1)B1OC(C)(C)C(C)(C)O1 |
C16H28BNO4 | |
MFCD03840345 | |
n-boc-1,2,5,6-tetrahydropyridine-4-boronic acid pinacol ester, tert-butyl 4-4,4,5,5-tetramethyl-1,3,2-dioxaborolan-2-yl-5,6-dihydropyridine-1 2h-carboxylate, 1-n-boc-4-4,4,5,5-tetramethyl-1,3,2 dioxaborolan-2-yl-3,6-dihydro-2h-pyridine, n-boc-1,2,3,6-tetrahydropyridine-4-boronic acid pinacol ester, tert-butyl 4-4,4,5,5-tetramethyl-1,3,2-dioxaborolan-2-yl-3,6-dihydro-2h-pyridine-1-carboxylate, tert-butyl 5,6-dihydro-4-4,4,5,5-tetramethyl-1,3,2-dioxaborolan-2-yl pyridine-1 2h-carboxylate, 3,6-dihydro-2h-pyridine-1-n-boc-4-boronic acid pinacol ester, n-tert-butoxycarbonyl-1,2,3,6-tetrahydropyridine-4-boronic acid pinacol ester, 1-boc-4-4,4,5,5-tetramethyl-1,3,2 dioxaborolan-2-yl-3,6-dihydro-2h-pyridine, tert-butyl 4-tetramethyl-1,3,2-dioxaborolan-2-yl-1,2,3,6-tetrahydropyridine-1-carboxylate | |
tert-butyl 4-(4,4,5,5-tetramethyl-1,3,2-dioxaborolan-2-yl)-3,6-dihydro-2H-pyridine-1-carboxylate |
Safety and Handling
TSCA : No
Recommended Storage : Ambient temperatures
RUO – Research Use Only