Please login to your online account to display your discounted pricing

Alfa Aesar™ N-Boc-1-(2,4-dinitrophenyl)-L-histidine, 99%

Chemical Identifiers

Chemical Name or Material N-Boc-1-(2,4-dinitrophenyl)-L-histidine
CAS 25024-53-7
Formula Weight 421.36
MDL Number MFCD00065966
Beilstein 771922
Synonym Boc-His(Dnp)-OH
View More Specs
Products
Catalog Number Mfr. No. Quantity Price Quantity    

AAH6342106

  SDS
alfa aesar™
H6342106
5g CALL N/A N/A
 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

AAH6342114

  SDS
alfa aesar™
H6342114
25g CALL N/A N/A
 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

AAH6342122

  SDS
alfa aesar™
H6342122
100g CALL N/A N/A
 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
Description & Specifications

Specifications

Formula Weight 421.36
MDL Number MFCD00065966
TSCA TSCA
Assay 99%
Sensitivity Light Sensitive
CAS 25024-53-7
Physical Form Solid
Synonym Boc-His(Dnp)-OH
Beilstein 771922
Chemical Name or Material N-Boc-1-(2,4-dinitrophenyl)-L-histidine
Quantity 5g

Applications
It is an important raw material and intermediate used in organic synthesis, pharmaceuticals, agrochemicals and dyestuff.

Solubility
Slightly soluble in water.

Notes
Light Sensitive, store in dark. Store in cool. Keep container tightly closed in a dry and well-ventilated place. Store away from oxidizing agent.