missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Alfa Aesar™ N-Boc-1-(2,4-dinitrophenyl)-L-histidine, 99%

Manufacturer:  Alfa Aesar™ H6342122

 View more versions of this product

Catalog No. AAH6342122



Description

Description

Applications
It is an important raw material and intermediate used in organic synthesis, pharmaceuticals, agrochemicals and dyestuff.

Solubility
Slightly soluble in water.

Notes
Light Sensitive, store in dark. Store in cool. Keep container tightly closed in a dry and well-ventilated place. Store away from oxidizing agent.
Specifications

Specifications

N-Boc-1-(2,4-dinitrophenyl)-L-histidine
C17H19N5O8
771922
KPGVUOQMOHGHEW-LBPRGKRZSA-N
421.366
421.36
99%
Light Sensitive
25024-53-7
MFCD00065966
boc-his dnp-oh, a-boc-n im-2,4-dinitrophenyl-l-histidine, n-tert-butoxycarbonyl-1-2,4-dinitrophenyl-l-histidine, l-histidine, n-1,1-dimethylethoxy carbonyl-1-2,4-dinitrophenyl, 2s-2-tert-butoxycarbonyl amino-3-1-2,4-dinitrophenyl imidazol-4-yl propanoic acid, pubchem18935, boc-his dnp-oh-ipa, boc-his dnp-oh isopropanol solvate, n-1,1-dimethylethoxy carbonyl-1-2,4-dinitrophenyl-l-histidine, boc-his dnp-oh tlc, n
CC(C)(C)OC(=O)NC(CC1=CN(C=N1)C2=C(C=C(C=C2)[N+](=O)[O-])[N+](=O)[O-])C(=O)O
90690
Solid
100g

Chemical Identifiers

25024-53-7
421.366
KPGVUOQMOHGHEW-LBPRGKRZSA-N
90690
C17H19N5O8
MFCD00065966
boc-his dnp-oh, a-boc-n im-2,4-dinitrophenyl-l-histidine, n-tert-butoxycarbonyl-1-2,4-dinitrophenyl-l-histidine, l-histidine, n-1,1-dimethylethoxy carbonyl-1-2,4-dinitrophenyl, 2s-2-tert-butoxycarbonyl amino-3-1-2,4-dinitrophenyl imidazol-4-yl propanoic acid, pubchem18935, boc-his dnp-oh-ipa, boc-his dnp-oh isopropanol solvate, n-1,1-dimethylethoxy carbonyl-1-2,4-dinitrophenyl-l-histidine, boc-his dnp-oh tlc, n
CC(C)(C)OC(=O)NC(CC1=CN(C=N1)C2=C(C=C(C=C2)[N+](=O)[O-])[N+](=O)[O-])C(=O)O
Safety and Handling

Safety and Handling

TSCA : TSCA

SDS Documents