Please login to your online account to display your discounted pricing

Alfa Aesar™ N-Benzyltrifluoromethanesulfonamide, 97%

$47.46 - $144.65

Chemical Identifiers

Chemical Name or Material N-Benzyltrifluoromethanesulfonamide
CAS 36457-58-6
Molecular Formula C8H8F3NO2S
Linear Formula C6H5CH2NHSO2CF3
Formula Weight 239.22
MDL Number MFCD00042074
Beilstein 2119124
Synonym N-Benzyltriflamide
View More Specs
Products
Catalog Number Mfr. No. Quantity Price Quantity    

AAL1670506

  SDS
alfa aesar™
L1670506
5g Each for $47.46

AAL1670514

  SDS
alfa aesar™
L1670514
25g Each for $144.65
Description & Specifications

Specifications

Formula Weight 239.22
Chemical Name or Material N-Benzyltrifluoromethanesulfonamide
Health Hazard 2 GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
MDL Number MFCD00042074
Health Hazard 1 Warning
Molecular Formula C8H8F3NO2S
Assay 97%
Synonym N-Benzyltriflamide
Health Hazard 3 GHS P Statement
P261-P280-P305+P351+P338-P304+P340-P405-P501a
Avoid breathing dust/fume/gas/mist/vapors/spray.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
IF INHALED: Remove to fresh air and keep at rest in a position comfortable for breathing.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.
Melting Point 44° to 46°C
Linear Formula C6H5CH2NHSO2CF3
Quantity 5g
CAS 36457-58-6
Beilstein 2119124
Safety and Handling

Warning

GHS P Statement
P261-P280-P305+P351+P338-P304+P340-P405-P501a
Avoid breathing dust/fume/gas/mist/vapors/spray.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
IF INHALED: Remove to fresh air and keep at rest in a position comfortable for breathing.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.

GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.