missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Alfa Aesar™ N-Benzylquininium chloride, 95%

Manufacturer:  Alfa Aesar™ H5342406

 View more versions of this product

Catalog No. AAH5342406



Specifications

Specifications

N-Benzylquininium chloride
200°C to 205°C
67174-25-8
MFCD00198105
JYDIJFKNXHPWBJ-PRJMIWRBSA-M
(R)-[(2S,4R,5S)-1-benzyl-5-ethenyl-1-azoniabicyclo[2.2.2]octan-2-yl]-(6-methoxyquinolin-4-yl)methanol;chloride
133611806
95%
−235° (c=1.5 in Water)
5g
C27H31ClN2O2
n-benzylquininiumchloride chiralphase-transfercatalyst
COC1=CC2=C(C=CN=C2C=C1)C(C3CC4CC[N+]3(CC4C=C)CC5=CC=CC=C5)O.[Cl-]
451.007
451.01

Chemical Identifiers

67174-25-8
451.007
JYDIJFKNXHPWBJ-PRJMIWRBSA-M
133611806
COC1=CC2=C(C=CN=C2C=C1)C(C3CC4CC[N+]3(CC4C=C)CC5=CC=CC=C5)O.[Cl-]
C27H31ClN2O2
MFCD00198105
n-benzylquininiumchloride chiralphase-transfercatalyst
(R)-[(2S,4R,5S)-1-benzyl-5-ethenyl-1-azoniabicyclo[2.2.2]octan-2-yl]-(6-methoxyquinolin-4-yl)methanol;chloride
SDS Documents