Please login to your online account to display your discounted pricing

N-(Benzyloxycarbonyloxy)succinimide, 98%, ACROS Organics™


Quantity 25g
Packaging Glass bottle

Manufacturer: thermo fisher scientific  211170250

 View more versions of this product

Catalog No. AC211170250

 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.


Description & Specifications

Chemical Identifiers

Physical Form Fine Crystalline Powder
Assay ≥97.5%
Formula Weight 249.22
Quantity 25g
Color White to off-white
Melting Point 77° to 82°C
CAS Max % 100.0
Chemical Name or Material N-(Benzyloxycarbonyloxy)succinimide
Infrared Spectrum Authentic
Assay Percent Range 98%
Molecular Formula C12H11NO5
CAS 13139-17-8
Appearance of Solution 10% soln. in acetone, clear colorless to almost colorless
Loss on Drying 0.25% max. (60°C, 2 hrs, 1mm Hg)
Packaging Glass bottle
MDL Number MFCD00005513
Synonym Z-ONSu
CAS Min % 97.5

Specifications

Chemical Name or Material N-(Benzyloxycarbonyloxy)succinimide
CAS 13139-17-8
CAS Min % 97.5
CAS Max % 100.0
Assay ≥97.5%
Assay Percent Range 98%
Molecular Formula C12H11NO5
Formula Weight 249.22
MDL Number MFCD00005513
Synonym Z-ONSu