Please login to your online account to display your discounted pricing

Alfa Aesar™ N-Benzyloxycarbonylglycine thioamide, 99%

$178.53 - $178.53

Chemical Identifiers

Chemical Name or Material N-Benzyloxycarbonylglycine thioamide
CAS 49548-40-5
Molecular Formula C10H12N2O2S
Formula Weight 224.28g/mol
MDL Number MFCD03452777
Synonym N-Cbz-Glycine thioamide; Z-Glycine thioamide
View More Specs
Products
Catalog Number Mfr. No. Quantity Price Quantity    

AAH2600503

 
alfa aesar™
H26005-03
1g Each for $178.53
Description & Specifications

Specifications

Melting Point 143° to 145°C
Quantity 1g
Chemical Name or Material N-Benzyloxycarbonylglycine thioamide
CAS 49548-40-5
Assay 99%
Molecular Formula C10H12N2O2S
Formula Weight 224.28g/mol
MDL Number MFCD03452777
Synonym N-Cbz-Glycine thioamide; Z-Glycine thioamide
Health Hazard 1 Warning
Health Hazard 2 GHS H Statement
H302
Harmful if swallowed.
Health Hazard 3 GHS P Statement
P280f
Wear protective clothing.
Safety and Handling

Warning

GHS P Statement
P280f
Wear protective clothing.

GHS H Statement
H302
Harmful if swallowed.