missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Please login to your online account to display your discounted pricing
N-Benzyloxycarbonylglycine methyl ester, 98%, Thermo Scientific Chemicals
Supplier: Thermo Scientific Chemicals H6285814
Description
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Specifications
N-Benzyloxycarbonylglycine methyl ester | |
22°C to 26°C | |
C11H13NO4 | |
MFCD00043631 | |
z-gly-ome, z-glycine methyl ester, methyl 2-benzyloxy carbonyl amino acetate, carbobenzyloxyglycine methyl ester, n-cbz-glycine methyl ester, methyl n-benzyloxycarbonylglycinate, cbz-glycine methyl ester, carbobenzoxyglycine methyl ester, methyl 2-benzyloxycarbonylamino acetate, glycine, n-phenylmethoxy carbonyl-, methyl ester | |
COC(=O)CNC(=O)OCC1=CC=CC=C1 | |
223.23 | |
223.23 |
1212-53-9 | |
147°C to 151°C | |
C6H5CH2OCONHCH2CO2CH3 | |
25 g | |
DZYBBBYFLOPVOL-UHFFFAOYSA-N | |
methyl 2-{[(benzyloxy)carbonyl]amino}acetate | |
71033 | |
98% |
Chemical Identifiers
1212-53-9 | |
223.23 | |
DZYBBBYFLOPVOL-UHFFFAOYSA-N | |
71033 |
C11H13NO4 | |
MFCD00043631 | |
z-gly-ome, z-glycine methyl ester, methyl 2-benzyloxy carbonyl amino acetate, carbobenzyloxyglycine methyl ester, n-cbz-glycine methyl ester, methyl n-benzyloxycarbonylglycinate, cbz-glycine methyl ester, carbobenzoxyglycine methyl ester, methyl 2-benzyloxycarbonylamino acetate, glycine, n-phenylmethoxy carbonyl-, methyl ester | |
COC(=O)CNC(=O)OCC1=CC=CC=C1 |
Safety and Handling
EINECSNumber : 214-922-8
TSCA : No
Recommended Storage : Ambient temperatures
RUO – Research Use Only