missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Alfa Aesar™ N-Benzyloxycarbonylglycinamide, 97%

$35.93 - $302.26

Chemical Identifiers

CAS 949-90-6
Molecular Formula C10H12N2O3
Molecular Weight (g/mol) 208.217
MDL Number MFCD00042825
InChI Key HQYMUNCIMNFLDT-UHFFFAOYSA-N
Synonym z-gly-nh2, z-glycineamide, z-glycine amide, benzyl n-carbamoylmethyl carbamate, benzyl n-2-amino-2-oxoethyl carbamate, benzyl carbamoylmethylcarbamate, n-carbamoylmethyl phenylmethoxy carboxamide, n-benzyloxycarbonylglycinamide, z-glycinamide, cbz-glyamide
PubChem CID 70366
IUPAC Name benzyl N-(2-amino-2-oxoethyl)carbamate
SMILES C1=CC=C(C=C1)COC(=O)NCC(=O)N
View More Specs

Products 3
Catalog Number Mfr. No. Quantity Price Quantity  
Catalog Number Mfr. No. Quantity Price Quantity  
AAH6208506 View Documents Alfa Aesar™
H6208506
5g Each for $35.93
 
AAH6208514 View Documents Alfa Aesar™
H6208514
25g Each for $99.15
 
AAH6208522 View Documents Alfa Aesar™
H6208522
100g Each for $302.26
 
Specifications

Chemical Identifiers

949-90-6
208.217
HQYMUNCIMNFLDT-UHFFFAOYSA-N
70366
C1=CC=C(C=C1)COC(=O)NCC(=O)N
C10H12N2O3
MFCD00042825
z-gly-nh2, z-glycineamide, z-glycine amide, benzyl n-carbamoylmethyl carbamate, benzyl n-2-amino-2-oxoethyl carbamate, benzyl carbamoylmethylcarbamate, n-carbamoylmethyl phenylmethoxy carboxamide, n-benzyloxycarbonylglycinamide, z-glycinamide, cbz-glyamide
benzyl N-(2-amino-2-oxoethyl)carbamate

Specifications

136°C to 139°C
949-90-6
C6H5CH2OCONHCH2CONH2
2054564
HQYMUNCIMNFLDT-UHFFFAOYSA-N
benzyl N-(2-amino-2-oxoethyl)carbamate
70366
97%
5g
C10H12N2O3
MFCD00042825
z-gly-nh2, z-glycineamide, z-glycine amide, benzyl n-carbamoylmethyl carbamate, benzyl n-2-amino-2-oxoethyl carbamate, benzyl carbamoylmethylcarbamate, n-carbamoylmethyl phenylmethoxy carboxamide, n-benzyloxycarbonylglycinamide, z-glycinamide, cbz-glyamide
C1=CC=C(C=C1)COC(=O)NCC(=O)N
208.217
208.22
N-Benzyloxycarbonylglycinamide