Please login to your online account to display your discounted pricing

Alfa Aesar™ N-Benzyloxycarbonyl-O-tert-butyl-L-tyrosine methyl ester, 95%

Chemical Identifiers

Chemical Name or Material N-Benzyloxycarbonyl-O-tert-butyl-L-tyrosine methyl ester
CAS 5068-29-1
Formula Weight 385.46
MDL Number MFCD00237370
Beilstein 2676239
Synonym Z-Tyr(tBu)-OMe
View More Specs
Products
Catalog Number Mfr. No. Quantity Price Quantity    

AAH6692503

 
alfa aesar™
H6692503
1g CALL N/A N/A
 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

AAH6692506

 
alfa aesar™
H6692506
5g CALL N/A N/A
 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

AAH6692514

 
alfa aesar™
H6692514
25g CALL N/A N/A
 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
Description & Specifications

Specifications

Formula Weight 385.46
Chemical Name or Material N-Benzyloxycarbonyl-O-tert-butyl-L-tyrosine methyl ester
MDL Number MFCD00237370
Physical Form Solid
CAS 5068-29-1
Beilstein 2676239
Synonym Z-Tyr(tBu)-OMe
Assay 95%
Quantity 1g
Storage Note 1 -30° to -10°C
Safety and Handling

-30° to -10°C