Please login to your online account to display your discounted pricing

Alfa Aesar™ N-Benzyloxycarbonyl-O-tert-butyl-L-threonine dicyclohexylammonium salt, 95%

Chemical Identifiers

Chemical Name or Material N-Benzyloxycarbonyl-O-tert-butyl-L-threonine dicyclohexylammonium salt
CAS 16966-07-7
Formula Weight 490.69
MDL Number MFCD00077043
Synonym Z-Thr(tBu)-OH.DCHA
View More Specs
Products
Catalog Number Mfr. No. Quantity Price Quantity    

AAH6678706

 
alfa aesar™
H6678706
5g CALL N/A N/A
 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

AAH6678714

 
alfa aesar™
H6678714
25g CALL N/A N/A
 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
Description & Specifications

Specifications

Formula Weight 490.69
CAS 16966-07-7
Chemical Name or Material N-Benzyloxycarbonyl-O-tert-butyl-L-threonine dicyclohexylammonium salt
Assay 95%
MDL Number MFCD00077043
Synonym Z-Thr(tBu)-OH.DCHA
Physical Form Solid
Storage Note 1 -30° to -10°C
Quantity 5g
Safety and Handling

-30° to -10°C