Please login to your online account to display your discounted pricing

Alfa Aesar™ N-Benzyloxycarbonyl-O-benzyl-L-tyrosine, 95%

Chemical Identifiers

Chemical Name or Material N-Benzyloxycarbonyl-O-benzyl-L-tyrosine
CAS 16677-29-5
Formula Weight 405.44
MDL Number MFCD00037819
Beilstein 3178475
Synonym Z-Tyr(Bzl)-OH
View More Specs
Products
Catalog Number Mfr. No. Quantity Price Quantity    

AAH6348703

 
alfa aesar™
H6348703
1g CALL N/A N/A
 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

AAH6348706

 
alfa aesar™
H6348706
5g CALL N/A N/A
 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
Description & Specifications

Specifications

Formula Weight 405.44
Chemical Name or Material N-Benzyloxycarbonyl-O-benzyl-L-tyrosine
Physical Form Solid
Assay 95%
MDL Number MFCD00037819
CAS 16677-29-5
Beilstein 3178475
Synonym Z-Tyr(Bzl)-OH
Quantity 1g
Storage Note 1 2° to 8°C
Safety and Handling

2° to 8°C