missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Alfa Aesar™ N-Benzyloxycarbonyl-O-benzyl-L-serine, 95%

$68.31 - $914.47

Chemical Identifiers

CAS 20806-43-3
Molecular Formula C18H19NO5
Molecular Weight (g/mol) 329.352
MDL Number MFCD03939889
InChI Key CYYRLHUAMWRBHC-INIZCTEOSA-N
Synonym cbz-ser bzl-oh, z-ser bzl-oh, n-benzyloxycarbonyl-o-benzyl-l-serine, carbobenzyloxy-o-benzyl-l-serine, 2s-3-benzyloxy-2-benzyloxy carbonyl amino propanoic acid, z-l-ser-obzl, ambotzzaa1185, pubchem19040, n-cbz-o-benzyl-l-ser-oh, n-benzyloxycarbonyl-o-benzylserine
PubChem CID 6993425
IUPAC Name (2S)-3-phenylmethoxy-2-(phenylmethoxycarbonylamino)propanoic acid
SMILES C1=CC=C(C=C1)COCC(C(=O)O)NC(=O)OCC2=CC=CC=C2
View More Specs

Products 3
Catalog Number Mfr. No. Quantity Price Quantity  
Catalog Number Mfr. No. Quantity Price Quantity  
AAH62748MD View Documents Alfa Aesar™
H62748MD
250mg Each for $68.31
 
AAH6274803 View Documents Alfa Aesar™
H6274803
1g Each for $197.39
 
AAH6274806 View Documents Alfa Aesar™
H6274806
5g Each for $914.47
 
Description

Description

Applications
N-Benzyloxycarbonyl-O-benzyl-L-serine is used as pharmaceutical intermediate.

Solubility
Slightly soluble in water.

Notes
Store in a cool, dry place, in a well sealed container. Store away from oxidizing agents.
Specifications

Chemical Identifiers

20806-43-3
329.352
CYYRLHUAMWRBHC-INIZCTEOSA-N
6993425
C1=CC=C(C=C1)COCC(C(=O)O)NC(=O)OCC2=CC=CC=C2
C18H19NO5
MFCD03939889
cbz-ser bzl-oh, z-ser bzl-oh, n-benzyloxycarbonyl-o-benzyl-l-serine, carbobenzyloxy-o-benzyl-l-serine, 2s-3-benzyloxy-2-benzyloxy carbonyl amino propanoic acid, z-l-ser-obzl, ambotzzaa1185, pubchem19040, n-cbz-o-benzyl-l-ser-oh, n-benzyloxycarbonyl-o-benzylserine
(2S)-3-phenylmethoxy-2-(phenylmethoxycarbonylamino)propanoic acid

Specifications

250mg
C18H19NO5
cbz-ser bzl-oh, z-ser bzl-oh, n-benzyloxycarbonyl-o-benzyl-l-serine, carbobenzyloxy-o-benzyl-l-serine, 2s-3-benzyloxy-2-benzyloxy carbonyl amino propanoic acid, z-l-ser-obzl, ambotzzaa1185, pubchem19040, n-cbz-o-benzyl-l-ser-oh, n-benzyloxycarbonyl-o-benzylserine
C1=CC=C(C=C1)COCC(C(=O)O)NC(=O)OCC2=CC=CC=C2
329.352
329.35
N-Benzyloxycarbonyl-O-benzyl-L-serine
20806-43-3
MFCD03939889
CYYRLHUAMWRBHC-INIZCTEOSA-N
(2S)-3-phenylmethoxy-2-(phenylmethoxycarbonylamino)propanoic acid
6993425
95%