Please login to your online account to display your discounted pricing

Alfa Aesar™ N-Benzyloxycarbonyl-O-benzyl-D-tyrosine, 95%

Chemical Identifiers

Chemical Name or Material N-Benzyloxycarbonyl-O-benzyl-D-tyrosine
CAS 92455-53-3
Formula Weight 405.45
MDL Number MFCD00153349
Beilstein 6941737
Synonym Z-D-Tyr(Bzl)-OH
View More Specs
Products
Catalog Number Mfr. No. Quantity Price Quantity    

AAH6604906

 
alfa aesar™
H6604906
5g CALL N/A N/A
 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

AAH6604914

 
alfa aesar™
H6604914
25g CALL N/A N/A
 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
Description & Specifications

Specifications

Melting Point 113° to 119°C
Formula Weight 405.45
Chemical Name or Material N-Benzyloxycarbonyl-O-benzyl-D-tyrosine
Assay 95%
CAS 92455-53-3
MDL Number MFCD00153349
Quantity 5g
Color White
Beilstein 6941737
Physical Form Powder
Storage Note 1 -30° to -10°C
Synonym Z-D-Tyr(Bzl)-OH
Safety and Handling

-30° to -10°C