missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Alfa Aesar™ N-Benzyloxycarbonyl-N-methyl-L-valine, 95%

$28.84 - $371.65

Chemical Identifiers

CAS 42417-65-2
Molecular Formula C14H19NO4
Molecular Weight (g/mol) 265.309
MDL Number MFCD00153397
InChI Key NNEHOKZDWLJKHP-LBPRGKRZSA-N
Synonym cbz-n-methyl-l-valine, a-methyl-l-valine, z-n-me-val-oh, cbz-n-me-val-oh, z-n-methyl-l-valine, s-2-benzyloxy carbonyl methyl amino-3-methylbutanoic acid, z-meval-oh, ambotzzaa1019, cbz-n-d-me-val-oh, cbz-n-methyl-l-val-oh, z-n-
PubChem CID 7016371
IUPAC Name (2S)-3-methyl-2-[methyl(phenylmethoxycarbonyl)amino]butanoic acid
SMILES CC(C)C(C(=O)O)N(C)C(=O)OCC1=CC=CC=C1
View More Specs

Products 3
Catalog Number Mfr. No. Quantity Price Quantity  
Catalog Number Mfr. No. Quantity Price Quantity  
AAH65485MD View Documents Alfa Aesar™
H65485MD
250mg Each for $28.84
 
AAH6548503 View Documents Alfa Aesar™
H6548503
1g Each for $81.73
 
AAH6548506 View Documents Alfa Aesar™
H6548506
5g Each for $371.65
 
Specifications

Chemical Identifiers

42417-65-2
265.309
NNEHOKZDWLJKHP-LBPRGKRZSA-N
7016371
CC(C)C(C(=O)O)N(C)C(=O)OCC1=CC=CC=C1
C14H19NO4
MFCD00153397
cbz-n-methyl-l-valine, a-methyl-l-valine, z-n-me-val-oh, cbz-n-me-val-oh, z-n-methyl-l-valine, s-2-benzyloxy carbonyl methyl amino-3-methylbutanoic acid, z-meval-oh, ambotzzaa1019, cbz-n-d-me-val-oh, cbz-n-methyl-l-val-oh, z-n-
(2S)-3-methyl-2-[methyl(phenylmethoxycarbonyl)amino]butanoic acid

Specifications

42417-65-2
MFCD00153397
cbz-n-methyl-l-valine, a-methyl-l-valine, z-n-me-val-oh, cbz-n-me-val-oh, z-n-methyl-l-valine, s-2-benzyloxy carbonyl methyl amino-3-methylbutanoic acid, z-meval-oh, ambotzzaa1019, cbz-n-d-me-val-oh, cbz-n-methyl-l-val-oh, z-n-
CC(C)C(C(=O)O)N(C)C(=O)OCC1=CC=CC=C1
265.309
265.31
95%
68°C to 70°C
N-Benzyloxycarbonyl-N-methyl-L-valine
C14H19NO4
1990143
NNEHOKZDWLJKHP-LBPRGKRZSA-N
(2S)-3-methyl-2-[methyl(phenylmethoxycarbonyl)amino]butanoic acid
7016371
Crystalline Solid
White
250mg