Please login to your online account to display your discounted pricing

Alfa Aesar™ N-Benzyloxycarbonyl-N-methyl-L-isoleucine, 95%

Chemical Identifiers

Chemical Name or Material N-Benzyloxycarbonyl-N-methyl-L-isoleucine
CAS 42417-66-3
Formula Weight 279.34
MDL Number MFCD00235876
Beilstein 6589752
Synonym Z-N-Me-Ile-OH; Z-N-Methyl-L-isoleucine
View More Specs
Products
Catalog Number Mfr. No. Quantity Price Quantity    

AAH6562503

 
alfa aesar™
H6562503
1g CALL N/A N/A
 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

AAH6562506

 
alfa aesar™
H6562506
5g CALL N/A N/A
 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

AAH65625MD

 
alfa aesar™
H65625MD
250mg CALL N/A N/A
 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
Description & Specifications

Specifications

Formula Weight 279.34
Chemical Name or Material N-Benzyloxycarbonyl-N-methyl-L-isoleucine
Physical Form Powder
CAS 42417-66-3
MDL Number MFCD00235876
Beilstein 6589752
Synonym Z-N-Me-Ile-OH; Z-N-Methyl-L-isoleucine
Color White to off-white
Quantity 1g
Storage Note 1 2° to 8°C
Assay 95%
Safety and Handling

2° to 8°C