missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Alfa Aesar™ N-Benzyloxycarbonyl-N-methyl-DL-alanine dicyclohexylammonium salt, 95%

$29.45 - $422.54

Chemical Identifiers

CAS 91738-83-9
Molecular Formula C24H38N2O4
Molecular Weight (g/mol) 418.578
MDL Number MFCD00672350
InChI Key RHGOEFBRIGKCFJ-UHFFFAOYSA-N
Synonym z-n-me-dl-ala-oh dcha, 2-benzyloxy carbonyl methyl amino propanoic acid; dicha, dicyclohexylamine 2-benzyloxy carbonyl methyl amino propanoate
PubChem CID 56777042
IUPAC Name N-cyclohexylcyclohexanamine;2-[methyl(phenylmethoxycarbonyl)amino]propanoic acid
SMILES CC(C(=O)O)N(C)C(=O)OCC1=CC=CC=C1.C1CCC(CC1)NC2CCCCC2
View More Specs

Products 3
Catalog Number Mfr. No. Quantity Price Quantity  
Catalog Number Mfr. No. Quantity Price Quantity  
AAH65851MD View Documents Alfa Aesar™
H65851MD
250mg Each for $29.45
 
AAH6585103 View Documents Alfa Aesar™
H6585103
1g Each for $92.68
 
AAH6585106 View Documents Alfa Aesar™
H6585106
5g Each for $422.54
 
Specifications

Chemical Identifiers

91738-83-9
418.578
RHGOEFBRIGKCFJ-UHFFFAOYSA-N
56777042
CC(C(=O)O)N(C)C(=O)OCC1=CC=CC=C1.C1CCC(CC1)NC2CCCCC2
C24H38N2O4
MFCD00672350
z-n-me-dl-ala-oh dcha, 2-benzyloxy carbonyl methyl amino propanoic acid; dicha, dicyclohexylamine 2-benzyloxy carbonyl methyl amino propanoate
N-cyclohexylcyclohexanamine;2-[methyl(phenylmethoxycarbonyl)amino]propanoic acid

Specifications

91738-83-9
MFCD00672350
RHGOEFBRIGKCFJ-UHFFFAOYSA-N
N-cyclohexylcyclohexanamine;2-[methyl(phenylmethoxycarbonyl)amino]propanoic acid
56777042
Solid
250mg
C24H38N2O4
z-n-me-dl-ala-oh dcha, 2-benzyloxy carbonyl methyl amino propanoic acid; dicha, dicyclohexylamine 2-benzyloxy carbonyl methyl amino propanoate
CC(C(=O)O)N(C)C(=O)OCC1=CC=CC=C1.C1CCC(CC1)NC2CCCCC2
418.578
418.58
95%
N-Benzyloxycarbonyl-N-methyl-DL-alanine dicyclohexylammonium salt
Safety and Handling

Safety and Handling

storageNote1 : 2° to 8°C