missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Please login to your online account to display your discounted pricing
N-Benzyloxycarbonyl-N-methyl-D-valine, 95%, Thermo Scientific™
Supplier: Thermo Scientific Chemicals H6553106
Description
This Thermo Scientific brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.
Specifications
N-Benzyloxycarbonyl-N-methyl-D-valine | |
White | |
MFCD04973293 | |
z-d-n-me-val-oh, n-cbz-n-methyl-d-valine, r-2-benzyloxy carbonyl methyl amino-3-methylbutanoic acid, ambotzzaa1004, benzyloxycarbonyl-n-alpha-methyl-d-valine, n-benzyloxy carbonyl-n-methyl-d-valine, d-valine, n-methyl-n-phenylmethoxy carbonyl, 2r-2-benzyloxy carbonyl methyl amino-3-methylbutanoic acid | |
CC(C)[C@@H](N(C)C(=O)OCC1=CC=CC=C1)C(O)=O | |
265.31 | |
265.31 | |
5 g |
53978-73-7 | |
C14H19NO4 | |
3593385 | |
NNEHOKZDWLJKHP-GFCCVEGCSA-N | |
(2R)-2-{[(benzyloxy)carbonyl](methyl)amino}-3-methylbutanoic acid | |
7018026 | |
95% | |
Solid |
Chemical Identifiers
53978-73-7 | |
265.31 | |
NNEHOKZDWLJKHP-GFCCVEGCSA-N | |
7018026 | |
CC(C)[C@@H](N(C)C(=O)OCC1=CC=CC=C1)C(O)=O |
C14H19NO4 | |
MFCD04973293 | |
z-d-n-me-val-oh, n-cbz-n-methyl-d-valine, r-2-benzyloxy carbonyl methyl amino-3-methylbutanoic acid, ambotzzaa1004, benzyloxycarbonyl-n-alpha-methyl-d-valine, n-benzyloxy carbonyl-n-methyl-d-valine, d-valine, n-methyl-n-phenylmethoxy carbonyl, 2r-2-benzyloxy carbonyl methyl amino-3-methylbutanoic acid | |
(2R)-2-{[(benzyloxy)carbonyl](methyl)amino}-3-methylbutanoic acid |
Safety and Handling
TSCA : No
storageNote1 : 2° to 8°C
Recommended Storage : Keep cold