Please login to your online account to display your discounted pricing

Alfa Aesar™ N-Benzyloxycarbonyl-N-methyl-D-alanine, 95%

Chemical Identifiers

Chemical Name or Material N-Benzyloxycarbonyl-N-methyl-D-alanine
CAS 68223-03-0
Formula Weight 237.26
MDL Number MFCD00153387
Beilstein 6807578
Synonym Z-N-Me-D-Ala-OH; Z-N-Methyl-D-alanine
View More Specs
Products
Catalog Number Mfr. No. Quantity Price Quantity    

AAH65685MD

 
alfa aesar™
H65685MD
250mg CALL N/A N/A
 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

AAH6568506

 
alfa aesar™
H6568506
5g CALL N/A N/A
 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

AAH6568503

 
alfa aesar™
H6568503
1g CALL N/A N/A
 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
Description & Specifications

Specifications

Melting Point 215° to 219°C
Formula Weight 237.26
MDL Number MFCD00153387
Chemical Name or Material N-Benzyloxycarbonyl-N-methyl-D-alanine
CAS 68223-03-0
Assay 95%
Synonym Z-N-Me-D-Ala-OH; Z-N-Methyl-D-alanine
Physical Form Solid
Beilstein 6807578
Quantity 250mg
Storage Note 1 2° to 8°C
Safety and Handling

2° to 8°C