missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Alfa Aesar™ N-Benzyloxycarbonyl-L-valylglycine ethyl ester, 97%

$87.23 - $346.24

Chemical Identifiers

CAS 2766-17-8
Molecular Formula C17H24N2O5
Molecular Weight (g/mol) 336.388
InChI Key FZTGKCOLBDXREB-HNNXBMFYSA-N
Synonym ethyl 2-2s-2-benzyloxy carbonyl amino-3-methylbutanamido acetate, z-val-gly-oet
PubChem CID 7015716
IUPAC Name ethyl 2-[[(2S)-3-methyl-2-(phenylmethoxycarbonylamino)butanoyl]amino]acetate
SMILES CCOC(=O)CNC(=O)C(C(C)C)NC(=O)OCC1=CC=CC=C1
View More Specs

Products 2
Catalog Number Mfr. No. Quantity Price Quantity  
Catalog Number Mfr. No. Quantity Price Quantity  
AAH6512603 View Documents Alfa Aesar™
H6512603
1g Each for $87.23
 
AAH6512606 View Documents Alfa Aesar™
H6512606
5g Each for $346.24
 
Specifications

Chemical Identifiers

2766-17-8
336.388
ethyl 2-2s-2-benzyloxy carbonyl amino-3-methylbutanamido acetate, z-val-gly-oet
ethyl 2-[[(2S)-3-methyl-2-(phenylmethoxycarbonylamino)butanoyl]amino]acetate
C17H24N2O5
FZTGKCOLBDXREB-HNNXBMFYSA-N
7015716
CCOC(=O)CNC(=O)C(C(C)C)NC(=O)OCC1=CC=CC=C1

Specifications

2766-17-8
1893763
FZTGKCOLBDXREB-HNNXBMFYSA-N
ethyl 2-[[(2S)-3-methyl-2-(phenylmethoxycarbonylamino)butanoyl]amino]acetate
7015716
Solid
1g
C17H24N2O5
ethyl 2-2s-2-benzyloxy carbonyl amino-3-methylbutanamido acetate, z-val-gly-oet
CCOC(=O)CNC(=O)C(C(C)C)NC(=O)OCC1=CC=CC=C1
336.388
336.39
97%
N-Benzyloxycarbonyl-L-valylglycine ethyl ester
Safety and Handling

Safety and Handling

TSCA : No

Recommended Storage : Ambient temperatures