missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Alfa Aesar™ N-Benzyloxycarbonyl-L-valylglycine ethyl ester, 97%

Manufacturer:  Alfa Aesar™ H6512606

 View more versions of this product

Catalog No. AAH6512606



Specifications

Specifications

N-Benzyloxycarbonyl-L-valylglycine ethyl ester
C17H24N2O5
ethyl 2-2s-2-benzyloxy carbonyl amino-3-methylbutanamido acetate, z-val-gly-oet
CCOC(=O)CNC(=O)C(C(C)C)NC(=O)OCC1=CC=CC=C1
336.388
336.39
97%
2766-17-8
1893763
FZTGKCOLBDXREB-HNNXBMFYSA-N
ethyl 2-[[(2S)-3-methyl-2-(phenylmethoxycarbonylamino)butanoyl]amino]acetate
7015716
Solid
5g

Chemical Identifiers

2766-17-8
336.388
ethyl 2-2s-2-benzyloxy carbonyl amino-3-methylbutanamido acetate, z-val-gly-oet
ethyl 2-[[(2S)-3-methyl-2-(phenylmethoxycarbonylamino)butanoyl]amino]acetate
C17H24N2O5
FZTGKCOLBDXREB-HNNXBMFYSA-N
7015716
CCOC(=O)CNC(=O)C(C(C)C)NC(=O)OCC1=CC=CC=C1
Safety and Handling

Safety and Handling

TSCA : No

Recommended Storage : Ambient temperatures

Documents