missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Alfa Aesar™ N-Benzyloxycarbonyl-L-valinamide, 97%

$124.00 - $505.81

Chemical Identifiers

CAS 13139-28-1
Molecular Formula C13H18N2O3
Molecular Weight (g/mol) 250.298
MDL Number MFCD00191170
InChI Key LWAHIZLDUQONHV-NSHDSACASA-N
Synonym z-val-nh2, z-l-valine amide, s-benzyl 1-amino-3-methyl-1-oxobutan-2-yl carbamate, benzyl n-1s-1-carbamoyl-2-methylpropyl carbamate, n-cbz-l-valinamide
PubChem CID 10562630
IUPAC Name benzyl N-[(2S)-1-amino-3-methyl-1-oxobutan-2-yl]carbamate
SMILES CC(C)C(C(=O)N)NC(=O)OCC1=CC=CC=C1
View More Specs

Products 2
Catalog Number Mfr. No. Quantity Price Quantity  
Catalog Number Mfr. No. Quantity Price Quantity  
AAH6600406 View Documents Alfa Aesar™
H6600406
5g Each for $124.00
 
AAH6600414 View Documents Alfa Aesar™
H6600414
25g Each for $505.81
 
Specifications

Chemical Identifiers

13139-28-1
250.298
LWAHIZLDUQONHV-NSHDSACASA-N
10562630
CC(C)C(C(=O)N)NC(=O)OCC1=CC=CC=C1
C13H18N2O3
MFCD00191170
z-val-nh2, z-l-valine amide, s-benzyl 1-amino-3-methyl-1-oxobutan-2-yl carbamate, benzyl n-1s-1-carbamoyl-2-methylpropyl carbamate, n-cbz-l-valinamide
benzyl N-[(2S)-1-amino-3-methyl-1-oxobutan-2-yl]carbamate

Specifications

5g
C13H18N2O3
z-val-nh2, z-l-valine amide, s-benzyl 1-amino-3-methyl-1-oxobutan-2-yl carbamate, benzyl n-1s-1-carbamoyl-2-methylpropyl carbamate, n-cbz-l-valinamide
CC(C)C(C(=O)N)NC(=O)OCC1=CC=CC=C1
250.298
250.3
97%
13139-28-1
MFCD00191170
LWAHIZLDUQONHV-NSHDSACASA-N
benzyl N-[(2S)-1-amino-3-methyl-1-oxobutan-2-yl]carbamate
10562630
Solid
N-Benzyloxycarbonyl-L-valinamide
Safety and Handling

Safety and Handling

storageNote1 : -30° to -10°C