missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Please login to your online account to display your discounted pricing
N-Benzyloxycarbonyl-L-tyrosine methyl ester, 98%, Thermo Scientific Chemicals
$88.22 - $1,215.22
Chemical Identifiers
CAS | 13512-31-7 |
---|---|
Molecular Formula | C18H19NO5 |
Molecular Weight (g/mol) | 329.35 |
MDL Number | MFCD00038784 |
InChI Key | SLLMDHBKALJDBW-XISACWJONA-N |
Synonym | z-tyr-ome, z-l-tyrosine methyl ester, n-cbz-l-tyrosine methyl ester, s-methyl 2-benzyloxy carbonyl amino-3-4-hydroxyphenyl propanoate, cbz-tyr-ome, z-l-tyr-ome, z-tyr-och3, cbz-tyrosine methyl ester, n-cbz-l-tyrosinemethylester, n-benzyloxycarbonyltyrosine methyl ester |
PubChem CID | 7010613 |
SMILES | COC(=O)[C@H](CC1=CC=C(O)C=C1)NC(=O)OCC1=CC=CC=C1 |
Catalog Number | Mfr. No. | Quantity | Price | Quantity | |||||
---|---|---|---|---|---|---|---|---|---|
Catalog Number | Mfr. No. | Quantity | Price | Quantity | |||||
AAH6311306
|
Thermo Scientific Chemicals
H6311306 |
5 g |
N/A
|
|
|||||
AAH6311314
|
Thermo Scientific Chemicals
H6311314 |
25 g |
N/A
|
|
|||||
AAH6311322
|
Thermo Scientific Chemicals
H6311322 |
100 g |
N/A
|
|
|||||
Description
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Chemical Identifiers
13512-31-7 | |
329.35 | |
SLLMDHBKALJDBW-XISACWJONA-N | |
7010613 |
C18H19NO5 | |
MFCD00038784 | |
z-tyr-ome, z-l-tyrosine methyl ester, n-cbz-l-tyrosine methyl ester, s-methyl 2-benzyloxy carbonyl amino-3-4-hydroxyphenyl propanoate, cbz-tyr-ome, z-l-tyr-ome, z-tyr-och3, cbz-tyrosine methyl ester, n-cbz-l-tyrosinemethylester, n-benzyloxycarbonyltyrosine methyl ester | |
COC(=O)[C@H](CC1=CC=C(O)C=C1)NC(=O)OCC1=CC=CC=C1 |
Specifications
93°C to 97°C (decomposition) | |
C18H19NO5 | |
z-tyr-ome, z-l-tyrosine methyl ester, n-cbz-l-tyrosine methyl ester, s-methyl 2-benzyloxy carbonyl amino-3-4-hydroxyphenyl propanoate, cbz-tyr-ome, z-l-tyr-ome, z-tyr-och3, cbz-tyrosine methyl ester, n-cbz-l-tyrosinemethylester, n-benzyloxycarbonyltyrosine methyl ester | |
COC(=O)[C@H](CC1=CC=C(O)C=C1)NC(=O)OCC1=CC=CC=C1 | |
329.35 | |
329.35 | |
5 g | |
N-Benzyloxycarbonyl-L-tyrosine methyl ester |
13512-31-7 | |
MFCD00038784 | |
SLLMDHBKALJDBW-XISACWJONA-N | |
methyl 2-{[(benzyloxy)carbonyl]amino}-3-(4-hydroxyphenyl)propanoate | |
7010613 | |
98% | |
Solid |
Safety and Handling
EINECSNumber : 426-320-4
TSCA : No
storageNote1 : 2° to 8°C
Recommended Storage : Keep cold
RUO – Research Use Only