missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Alfa Aesar™ N-Benzyloxycarbonyl-L-tryptophan benzyl ester, 95%

$61.54 - $245.20

Chemical Identifiers

CAS 69876-37-5
Molecular Formula C26H24N2O4
Molecular Weight (g/mol) 428.488
MDL Number MFCD00191163
InChI Key UHYCVEDLXBEKPC-DEOSSOPVSA-N
Synonym z-trp-obzl, nalpha-cbz-l-tryptophan benzyl ester, benzyl 2s-2-benzyloxy carbonyl amino-3-1h-indol-3-yl propanoate, z-l-trp-obzl, s-benzyl 2-benzyloxycarbonylamino-3-1h-indol-3-yl propanoate, benzyl 2s-2-benzyloxycarbonylamino-3-1h-indol-3-yl propanoate
PubChem CID 14102143
IUPAC Name benzyl (2S)-3-(1H-indol-3-yl)-2-(phenylmethoxycarbonylamino)propanoate
SMILES C1=CC=C(C=C1)COC(=O)C(CC2=CNC3=CC=CC=C32)NC(=O)OCC4=CC=CC=C4
View More Specs

Products 2
Catalog Number Mfr. No. Quantity Price Quantity  
Catalog Number Mfr. No. Quantity Price Quantity  
AAH6570203 View Documents Alfa Aesar™
H6570203
1g Each for $61.54
 
AAH6570206 View Documents Alfa Aesar™
H6570206
5g Each for $245.20
 
Specifications

Chemical Identifiers

69876-37-5
428.488
UHYCVEDLXBEKPC-DEOSSOPVSA-N
14102143
C1=CC=C(C=C1)COC(=O)C(CC2=CNC3=CC=CC=C32)NC(=O)OCC4=CC=CC=C4
C26H24N2O4
MFCD00191163
z-trp-obzl, nalpha-cbz-l-tryptophan benzyl ester, benzyl 2s-2-benzyloxy carbonyl amino-3-1h-indol-3-yl propanoate, z-l-trp-obzl, s-benzyl 2-benzyloxycarbonylamino-3-1h-indol-3-yl propanoate, benzyl 2s-2-benzyloxycarbonylamino-3-1h-indol-3-yl propanoate
benzyl (2S)-3-(1H-indol-3-yl)-2-(phenylmethoxycarbonylamino)propanoate

Specifications

69876-37-5
MFCD00191163
z-trp-obzl, nalpha-cbz-l-tryptophan benzyl ester, benzyl 2s-2-benzyloxy carbonyl amino-3-1h-indol-3-yl propanoate, z-l-trp-obzl, s-benzyl 2-benzyloxycarbonylamino-3-1h-indol-3-yl propanoate, benzyl 2s-2-benzyloxycarbonylamino-3-1h-indol-3-yl propanoate
C1=CC=C(C=C1)COC(=O)C(CC2=CNC3=CC=CC=C32)NC(=O)OCC4=CC=CC=C4
428.488
428.49
95%
1g
C26H24N2O4
4725277
UHYCVEDLXBEKPC-DEOSSOPVSA-N
benzyl (2S)-3-(1H-indol-3-yl)-2-(phenylmethoxycarbonylamino)propanoate
14102143
Powder
White
N-Benzyloxycarbonyl-L-tryptophan benzyl ester
Safety and Handling

Safety and Handling

storageNote1 : 2° to 8°C