Please login to your online account to display your discounted pricing

Alfa Aesar™ N-Benzyloxycarbonyl-L-threonylglycine ethyl ester, 97%

Chemical Identifiers

Chemical Name or Material N-Benzyloxycarbonyl-L-threonylglycine ethyl ester
CAS 27482-74-2
Formula Weight 338.36
MDL Number MFCD00191160
Beilstein 5131540
Synonym Z-Thr-Gly-OEt
View More Specs
Products
Catalog Number Mfr. No. Quantity Price Quantity    

AAH6512803

 
alfa aesar™
H6512803
1g CALL N/A N/A
 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

AAH6512806

 
alfa aesar™
H6512806
5g CALL N/A N/A
 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
Description & Specifications

Specifications

Formula Weight 338.36
Chemical Name or Material N-Benzyloxycarbonyl-L-threonylglycine ethyl ester
MDL Number MFCD00191160
CAS 27482-74-2
Assay 97%
Synonym Z-Thr-Gly-OEt
Beilstein 5131540
Quantity 1g
Physical Form Solid