Please login to your online account to display your discounted pricing

Alfa Aesar™ N-Benzyloxycarbonyl-L-threonine methyl ester, 97%

Chemical Identifiers

Chemical Name or Material N-Benzyloxycarbonyl-L-threonine methyl ester
CAS 57224-63-2
Formula Weight 267.28
MDL Number MFCD00038585
Beilstein 2220800
Synonym Z-Thr-OMe
View More Specs
Products
Catalog Number Mfr. No. Quantity Price Quantity    

AAH6637606

 
alfa aesar™
H6637606
5g CALL N/A N/A
 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

AAH6637622

 
alfa aesar™
H6637622
100g CALL N/A N/A
 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

AAH6637614

 
alfa aesar™
H6637614
25g CALL N/A N/A
 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
Description & Specifications

Specifications

Formula Weight 267.28
Melting Point 92° to 94°C
Chemical Name or Material N-Benzyloxycarbonyl-L-threonine methyl ester
CAS 57224-63-2
Physical Form Powder
MDL Number MFCD00038585
Beilstein 2220800
Synonym Z-Thr-OMe
Color White
Quantity 5g
Storage Note 1 -30° to -10°C
Assay 97%
Safety and Handling

-30° to -10°C