Please login to your online account to display your discounted pricing

Alfa Aesar™ N-Benzyloxycarbonyl-L-threonine benzyl ester, 99%

$141.64 - $141.64

Chemical Identifiers

Chemical Name or Material N-Benzyloxycarbonyl-L-threonine benzyl ester
CAS 16597-50-5
Molecular Formula C19H21NO5
Formula Weight 343.38
MDL Number MFCD00077041
Synonym N-Cbz-L-Threonine benzyl ester; Z-Thr-OBzl
View More Specs
Products
Catalog Number Mfr. No. Quantity Price Quantity    

AAB2267606

  SDS
alfa aesar™
B2267606
5g Each for $141.64
Description & Specifications

Specifications

Formula Weight 343.38
Chemical Name or Material N-Benzyloxycarbonyl-L-threonine benzyl ester
Melting Point 78° to 82°C
Quantity 5g
CAS 16597-50-5
Assay 99%
MDL Number MFCD00077041
Synonym N-Cbz-L-Threonine benzyl ester; Z-Thr-OBzl
Molecular Formula C19H21NO5