Please login to your online account to display your discounted pricing

Alfa Aesar™ N-Benzyloxycarbonyl-L-threoninamide, 97%

Chemical Identifiers

Chemical Name or Material N-Benzyloxycarbonyl-L-threoninamide
CAS 49705-98-8
Formula Weight 252.27
MDL Number MFCD00038587
Beilstein 2335408
Synonym Z-Thr-NH2
View More Specs
Products
Catalog Number Mfr. No. Quantity Price Quantity    

AAH6557006

 
alfa aesar™
H6557006
5g CALL N/A N/A
 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

AAH6557003

 
alfa aesar™
H6557003
1g CALL N/A N/A
 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

AAH65570MD

 
alfa aesar™
H65570MD
250mg CALL N/A N/A
 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
Description & Specifications

Specifications

Formula Weight 252.27
CAS 49705-98-8
MDL Number MFCD00038587
Assay 97%
Beilstein 2335408
Physical Form Powder
Synonym Z-Thr-NH2
Color White
Quantity 5g
Storage Note 1 -30° to -10°C
Chemical Name or Material N-Benzyloxycarbonyl-L-threoninamide
Safety and Handling

-30° to -10°C