Please login to your online account to display your discounted pricing

Alfa Aesar™ N-Benzyloxycarbonyl-L-tert-leucine, 99%

$194.38 - $497.23

Chemical Identifiers

Chemical Name or Material N-Benzyloxycarbonyl-L-tert-leucine
CAS 62965-10-0
Molecular Formula C14H19NO4
Formula Weight 265.3
MDL Number MFCD00137411
Synonym Z-tert-Leu-OH
View More Specs
Products
Catalog Number Mfr. No. Quantity Price Quantity    

AAH27132MD

  SDS
alfa aesar™
H27132MD
250mg Each for $194.38

AAH2713203

  SDS
alfa aesar™
H2713203
1g Each for $497.23
Description & Specifications

Specifications

Formula Weight 265.3
Chemical Name or Material N-Benzyloxycarbonyl-L-tert-leucine
Assay 99%
Molecular Formula C14H19NO4
MDL Number MFCD00137411
CAS 62965-10-0
Quantity 250mg
Synonym Z-tert-Leu-OH
Optical Rotation −4° (c=1 in Methanol)