Please login to your online account to display your discounted pricing

Alfa Aesar™ N-Benzyloxycarbonyl-L-serylglycine ethyl ester, 95%

Chemical Identifiers

Chemical Name or Material N-Benzyloxycarbonyl-L-serylglycine ethyl ester
CAS 4526-93-6
Formula Weight 324.33
MDL Number MFCD00191150
Beilstein 1893538
Synonym Z-Ser-Gly-OEt; Z-Seryl-L-glycine ethyl ester
View More Specs
Products
Catalog Number Mfr. No. Quantity Price Quantity    

AAH6532803

 
alfa aesar™
H6532803
1g CALL N/A N/A
 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

AAH65328MD

 
alfa aesar™
H65328MD
250mg CALL N/A N/A
 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

AAH6532806

 
alfa aesar™
H6532806
5g CALL N/A N/A
 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
Description & Specifications

Specifications

Formula Weight 324.33
Chemical Name or Material N-Benzyloxycarbonyl-L-serylglycine ethyl ester
MDL Number MFCD00191150
CAS 4526-93-6
Physical Form Solid
Assay 95%
Quantity 1g
Beilstein 1893538
Synonym Z-Ser-Gly-OEt; Z-Seryl-L-glycine ethyl ester
Storage Note 1 2° to 8°C
Safety and Handling

2° to 8°C