Please login to your online account to display your discounted pricing

Alfa Aesar™ N-Benzyloxycarbonyl-L-pyroglutamic acid N-succinimidyl ester, 95%

Chemical Identifiers

Chemical Name or Material N-Benzyloxycarbonyl-L-pyroglutamic acid N-succinimidyl ester
CAS 40291-26-7
Formula Weight 360.32
MDL Number MFCD00191144
Beilstein 5157479
Synonym Z-Pyr-Osu; N-Benzyloxycarbonyl-L-pyroglutamic acid N-hydroxysuccinimide ester
View More Specs
Products
Catalog Number Mfr. No. Quantity Price Quantity    

AAH6685803

 
alfa aesar™
H6685803
1g CALL N/A N/A
 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

AAH6685806

 
alfa aesar™
H6685806
5g CALL N/A N/A
 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

AAH6685814

 
alfa aesar™
H6685814
25g CALL N/A N/A
 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
Description & Specifications

Specifications

Formula Weight 360.32
Chemical Name or Material N-Benzyloxycarbonyl-L-pyroglutamic acid N-succinimidyl ester
Assay 95%
MDL Number MFCD00191144
CAS 40291-26-7
Physical Form Solid
Beilstein 5157479
Quantity 1g
Storage Note 1 -30° to -10°C
Synonym Z-Pyr-Osu; N-Benzyloxycarbonyl-L-pyroglutamic acid N-hydroxysuccinimide ester
Safety and Handling

-30° to -10°C