Please login to your online account to display your discounted pricing

Alfa Aesar™ N-Benzyloxycarbonyl-L-phenylalaninol, 98%

$128.07 - $1,710.14

Chemical Identifiers

Chemical Name or Material N-Benzyloxycarbonyl-L-phenylalaninol
CAS 6372-14-1
Molecular Formula C17H19NO3
Formula Weight 285.35
MDL Number MFCD00191138
Synonym (S)-2-Carbobenzyloxyamino-3-phenyl-1-propanol; N-Cbz-L-phenylalaninol
View More Specs
Products
Catalog Number Mfr. No. Quantity Price Quantity    

AAB2503606

 
alfa aesar™
B2503606
5g Each for $128.07

AAB2503614

 
alfa aesar™
B2503614
25g Each for $507.96

AAB2503622

 
alfa aesar™
B2503622
100g Each for $1,710.14
Description & Specifications

Specifications

Formula Weight 285.35
Chemical Name or Material N-Benzyloxycarbonyl-L-phenylalaninol
Assay 98%
Molecular Formula C17H19NO3
Melting Point 92° to 95°C
MDL Number MFCD00191138
CAS 6372-14-1
Quantity 5g
Synonym (S)-2-Carbobenzyloxyamino-3-phenyl-1-propanol; N-Cbz-L-phenylalaninol