Please login to your online account to display your discounted pricing

Alfa Aesar™ N-Benzyloxycarbonyl-L-phenylalanine N-succinimidyl ester, 95%

Chemical Identifiers

Chemical Name or Material N-Benzyloxycarbonyl-L-phenylalanine N-succinimidyl ester
CAS 3397-32-8
Formula Weight 396.4
MDL Number MFCD00053549
Beilstein 1557534
Synonym Z-Phe-Osu; N-Benzyloxycarbonyl-L-phenylalanine N-hydroxysuccinimide ester
View More Specs
Products
Catalog Number Mfr. No. Quantity Price Quantity    

AAH6522706

  SDS
alfa aesar™
H6522706
5g CALL N/A N/A
 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

AAH6522714

  SDS
alfa aesar™
H6522714
25g CALL N/A N/A
 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

AAH6522722

  SDS
alfa aesar™
H6522722
100g CALL N/A N/A
 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
Description & Specifications

Specifications

Formula Weight 396.4
Chemical Name or Material N-Benzyloxycarbonyl-L-phenylalanine N-succinimidyl ester
Physical Form Solid
Assay 95%
CAS 3397-32-8
Synonym Z-Phe-Osu; N-Benzyloxycarbonyl-L-phenylalanine N-hydroxysuccinimide ester
Storage Note 1 2° to 8°C
Quantity 5g
Beilstein 1557534
MDL Number MFCD00053549
Safety and Handling

2° to 8°C