Please login to your online account to display your discounted pricing

Alfa Aesar™ N-Benzyloxycarbonyl-L-phenylalaninamide, 95%

Chemical Identifiers

Chemical Name or Material N-Benzyloxycarbonyl-L-phenylalaninamide
CAS 4801-80-3
Formula Weight 298.34
MDL Number MFCD00077028
Synonym Z-Phe-NH2; Z-L-Phenylalaninamide
View More Specs
Products
Catalog Number Mfr. No. Quantity Price Quantity    

AAH6503406

 
alfa aesar™
H6503406
5g CALL N/A N/A
 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

AAH6503414

 
alfa aesar™
H6503414
25g CALL N/A N/A
 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

AAH6503422

 
alfa aesar™
H6503422
100g CALL N/A N/A
 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
Description & Specifications

Specifications

Formula Weight 298.34
Chemical Name or Material N-Benzyloxycarbonyl-L-phenylalaninamide
MDL Number MFCD00077028
CAS 4801-80-3
Synonym Z-Phe-NH2; Z-L-Phenylalaninamide
Assay 95%
Physical Form Solid
Quantity 5g
Storage Note 1 2° to 8°C
Safety and Handling

2° to 8°C