Please login to your online account to display your discounted pricing

Alfa Aesar™ N-Benzyloxycarbonyl-L-norleucine, 98+%

$41.30 - $41.30

Chemical Identifiers

Chemical Name or Material N-Benzyloxycarbonyl-L-norleucine
CAS 39608-30-5
Molecular Formula C14H19NO4
Formula Weight 265.3
MDL Number MFCD00063170
Beilstein 3210973
Synonym N-Cbz-L-norleucine; Z-Nle-OH
View More Specs
Products
Catalog Number Mfr. No. Quantity Price Quantity    

AAL09046MD

 
alfa aesar™
L09046MD
250mg Each for $41.30
Description & Specifications

Specifications

Formula Weight 265.3
Chemical Name or Material N-Benzyloxycarbonyl-L-norleucine
Assay ≥98%
Molecular Formula C14H19NO4
MDL Number MFCD00063170
CAS 39608-30-5
Beilstein 3210973
Quantity 250mg
Synonym N-Cbz-L-norleucine; Z-Nle-OH
Optical Rotation −10° (c=1 in Methanol)
Recommended Storage 2° to 8°C
Melting Point 56° to 58°C
Safety and Handling

2° to 8°C