missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Alfa Aesar™ N-Benzyloxycarbonyl-L-methionine, 98+%

$52.49 - $52.49

Chemical Identifiers

CAS 1152-62-1
Molecular Formula C13H17NO4S
Formula Weight 283.35
MDL Number MFCD00065124
Beilstein 2058696
Synonym N-Cbz-L-methionine; Z-Met-OH
Chemical Name or Material N-Benzyloxycarbonyl-L-methionine
View More Specs


Products ${productFamilyLength}
Catalog Number Mfr. No. Quantity Price Quantity  
Catalog Number Mfr. No. Quantity Price Quantity  
AAL0884006 SDS Alfa Aesar™
L0884006
5g Each for $52.49
Specifications Description & Specifications

Chemical Identifiers

1152-62-1
283.35
2058696
N-Benzyloxycarbonyl-L-methionine
C13H17NO4S
MFCD00065124
N-Cbz-L-methionine; Z-Met-OH

Specifications

1152-62-1
283.35
MFCD00065124
5g
−26° (c=1 in Methanol)
≥98%
C13H17NO4S
67° to 71°C
2058696
N-Cbz-L-methionine; Z-Met-OH
N-Benzyloxycarbonyl-L-methionine

Safety and Handling

Recommended Storage : 2° to 8°C