Please login to your online account to display your discounted pricing

Alfa Aesar™ N-Benzyloxycarbonyl-L-leucine, 98% (dry wt.), may cont. up to ca 5% solvent

$38.12 - $123.70

Chemical Identifiers

Chemical Name or Material N-Benzyloxycarbonyl-L-leucine
CAS 2018-66-8
Molecular Formula C14H19NO4
Formula Weight 265.3
MDL Number MFCD00026494
Beilstein 1253861
Synonym N-Cbz-L-leucine; Z-Leu-OH
View More Specs
Products
Catalog Number Mfr. No. Quantity Price Quantity    

AAL0847106

  SDS
alfa aesar™
L0847106
5g Each for $38.12

AAL0847114

  SDS
alfa aesar™
L0847114
25g Each for $123.70
Description & Specifications

Specifications

Formula Weight 265.3
Molecular Formula C14H19NO4
Chemical Name or Material N-Benzyloxycarbonyl-L-leucine
MDL Number MFCD00026494
Synonym N-Cbz-L-leucine; Z-Leu-OH
CAS 2018-66-8
Assay ≥98%
Quantity 5g
RTECS Number OH2921000
Beilstein 1253861
Assay Percent Notes (dry wt.), may cont. up to ≈5% solvent
Flash Point >110°C (230°F)
Optical Rotation −17.5° (c=2 in Ethanol)
Odor Odorless
Recommended Storage 2° to 8°C

Applications
It is employed as a reactant for peptide-based preparation of potential antitumor agents, preparation of cyclopropyl peptidomimetics and peptide synthesis.

Solubility
Not miscible or difficult to mix in water. Soluble in chloroform, DMSO and methanol.

Notes
Store away from oxidizing agents. Keep the container tightly closed and place it in a cool, dry and well ventilated condition. Protect from heat. Keep it refrigerated.

Safety and Handling

2° to 8°C