Please login to your online account to display your discounted pricing

Alfa Aesar™ N-Benzyloxycarbonyl-L-isoleucine dicyclohexylammonium salt, 98%

Chemical Identifiers

Chemical Name or Material N-Benzyloxycarbonyl-L-isoleucine dicyclohexylammonium salt
CAS 26699-00-3
Formula Weight 446.63
MDL Number MFCD00043243
Beilstein 6465170
Synonym Z-Ile-OH.DCHA
View More Specs
Products
Catalog Number Mfr. No. Quantity Price Quantity    

AAH6673314

 
alfa aesar™
H6673314
25g CALL N/A N/A
 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

AAH6673322

 
alfa aesar™
H6673322
100g CALL N/A N/A
 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
Description & Specifications

Specifications

Formula Weight 446.63
Melting Point 159° to 163°C
MDL Number MFCD00043243
Chemical Name or Material N-Benzyloxycarbonyl-L-isoleucine dicyclohexylammonium salt
Assay 98%
Synonym Z-Ile-OH.DCHA
Physical Form Powder
Beilstein 6465170
Storage Note 1 2° to 8°C
Color Off-white to white
Quantity 25g
CAS 26699-00-3
Safety and Handling

2° to 8°C