Please login to your online account to display your discounted pricing

Alfa Aesar™ N-Benzyloxycarbonyl-L-glutaminylglycine, 98%

$60.27 - $60.27

Chemical Identifiers

Chemical Name or Material N-Benzyloxycarbonyl-L-glutaminylglycine
CAS 6610-42-0
Formula Weight 337.33
MDL Number MFCD00055926
Beilstein 2179068
Synonym Z-Gln-Gly-OH
View More Specs
Products
Catalog Number Mfr. No. Quantity Price Quantity    

AAH6508803

 
alfa aesar™
H6508803
1g CALL N/A N/A
 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

AAH6508806

 
alfa aesar™
H6508806
5g CALL N/A N/A
 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

AAH65088MD

 
alfa aesar™
H65088MD
250mg Each for $60.27
Description & Specifications

Specifications

Formula Weight 337.33
CAS 6610-42-0
Chemical Name or Material N-Benzyloxycarbonyl-L-glutaminylglycine
MDL Number MFCD00055926
Beilstein 2179068
Assay 98%
Quantity 1g
Synonym Z-Gln-Gly-OH
Storage Note 1 -30° to -10°C
Physical Form Solid
Safety and Handling

-30° to -10°C