Please login to your online account to display your discounted pricing

N-Benzyloxycarbonyl-L-glutamic acid γ-tert-butyl ester, +98%, ACROS Organics™


Quantity 1g
Packaging Glass bottle

Manufacturer: thermo fisher scientific  AC426150010

 View more versions of this product

Catalog No. AC426150010

 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.


Description & Specifications

Chemical Identifiers

Formula Weight 337.37
Quantity 1g
CAS 725591
CAS Min % 98.0
CAS Max % 100.0
Chemical Name or Material N-Benzyloxycarbonyl-L-glutamic acid γ-tert-butyl ester
Packaging Glass bottle
Assay Percent Range 98+%
Molecular Formula C17H23NO6
MDL Number MFCD00038274

Specifications

Chemical Name or Material N-Benzyloxycarbonyl-L-glutamic acid γ-tert-butyl ester
CAS 725591
CAS Min % 98.0
CAS Max % 100.0
Assay Percent Range 98+%
Molecular Formula C17H23NO6
Formula Weight 337.37
MDL Number MFCD00038274