Please login to your online account to display your discounted pricing

Alfa Aesar™ N-Benzyloxycarbonyl-L-glutamic acid, 99%

$58.81 - $184.72

Chemical Identifiers

Chemical Name or Material N-Benzyloxycarbonyl-L-glutamic acid
CAS 1155-62-0
Molecular Formula C13H15NO6
Formula Weight 281.27
MDL Number MFCD00002801
Beilstein 2061272
Synonym N-Cbz-L-glutamic acid; Z-Glu-OH
View More Specs
Products
Catalog Number Mfr. No. Quantity Price Quantity    

AAA1768609

 
alfa aesar™
A1768609
10g Each for $58.81

AAA1768618

 
alfa aesar™
A1768618
50g Each for $184.72
Description & Specifications

Specifications

Formula Weight 281.27
Chemical Name or Material N-Benzyloxycarbonyl-L-glutamic acid
TSCA TSCA
Assay 99%
Molecular Formula C13H15NO6
Melting Point 108° to 116°C
MDL Number MFCD00002801
CAS 1155-62-0
Beilstein 2061272
Quantity 10g
Synonym N-Cbz-L-glutamic acid; Z-Glu-OH
Optical Rotation −7.5° (c=4 in Acetic Acid)