missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Alfa Aesar™ N-Benzyloxycarbonyl-L-glutamic acid 5-benzyl ester, 95%

$82.96 - $326.93

Chemical Identifiers

CAS 5680-86-4
Molecular Formula C20H21NO6
Molecular Weight (g/mol) 371.389
MDL Number MFCD00065697
InChI Key TWIVXHQQTRSWGO-KRWDZBQOSA-N
Synonym z-glu obzl-oh, cbz-glu obzl-oh, 2s-5-benzyloxy-2-benzyloxy carbonyl amino-5-oxopentanoic acid, 5-benzyl n-carbobenzoxy-l-glutamate, n-cbz-l-glutamic acid 5-benzyl ester, s-2-benzyloxycarbonylamino-pentanedioic acid 5-benzyl ester, s-5-benzyloxy-2-benzyloxy carbonyl amino-5-oxopentanoic acid, z-glu obzl oh, pubchem14964, z-glu-obzl-oh
PubChem CID 3049860
IUPAC Name (2S)-5-oxo-5-phenylmethoxy-2-(phenylmethoxycarbonylamino)pentanoic acid
SMILES C1=CC=C(C=C1)COC(=O)CCC(C(=O)O)NC(=O)OCC2=CC=CC=C2
View More Specs

Products 2
Catalog Number Mfr. No. Quantity Price Quantity  
Catalog Number Mfr. No. Quantity Price Quantity  
AAH6218106 View Documents Alfa Aesar™
H6218106
5g Each for $82.96
 
AAH6218114 View Documents Alfa Aesar™
H6218114
25g Each for $326.93
 
Description

Description

Applications
N-Benzyloxycarbonyl-L-glutamic acid 5-benzyl ester is used as pharmaceutical intermediates.

Solubility
Soluble in ethanol. Insoluble in water.

Notes
Keep container tightly closed. Store away from oxidizing agents.
Specifications

Chemical Identifiers

5680-86-4
371.389
TWIVXHQQTRSWGO-KRWDZBQOSA-N
3049860
C1=CC=C(C=C1)COC(=O)CCC(C(=O)O)NC(=O)OCC2=CC=CC=C2
C20H21NO6
MFCD00065697
z-glu obzl-oh, cbz-glu obzl-oh, 2s-5-benzyloxy-2-benzyloxy carbonyl amino-5-oxopentanoic acid, 5-benzyl n-carbobenzoxy-l-glutamate, n-cbz-l-glutamic acid 5-benzyl ester, s-2-benzyloxycarbonylamino-pentanedioic acid 5-benzyl ester, s-5-benzyloxy-2-benzyloxy carbonyl amino-5-oxopentanoic acid, z-glu obzl oh, pubchem14964, z-glu-obzl-oh
(2S)-5-oxo-5-phenylmethoxy-2-(phenylmethoxycarbonylamino)pentanoic acid

Specifications

5g
C20H21NO6
z-glu obzl-oh, cbz-glu obzl-oh, 2s-5-benzyloxy-2-benzyloxy carbonyl amino-5-oxopentanoic acid, 5-benzyl n-carbobenzoxy-l-glutamate, n-cbz-l-glutamic acid 5-benzyl ester, s-2-benzyloxycarbonylamino-pentanedioic acid 5-benzyl ester, s-5-benzyloxy-2-benzyloxy carbonyl amino-5-oxopentanoic acid, z-glu obzl oh, pubchem14964, z-glu-obzl-oh
C1=CC=C(C=C1)COC(=O)CCC(C(=O)O)NC(=O)OCC2=CC=CC=C2
371.389
371.38
N-Benzyloxycarbonyl-L-glutamic acid 5-benzyl ester
5680-86-4
MFCD00065697
TWIVXHQQTRSWGO-KRWDZBQOSA-N
(2S)-5-oxo-5-phenylmethoxy-2-(phenylmethoxycarbonylamino)pentanoic acid
3049860
95%